missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAF60C Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA529436
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Recommended positive controls: PC-3, U87-MG, SK-N-SH, NIH-3T3. Predicted reactivity: Mouse (100%), Rat (99%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BAF60C is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined.
Spécification
| BAF60C | |
| Polyclonal | |
| Unconjugated | |
| SMARCD3 | |
| 1500001J14Rik; 2210409C08Rik; 60 kDa BRG-1/Brm-associated factor subunit C; BAF60C; BRG1-associated factor 60C; chromatin remodeling complex BAF60C subunit; CRACD3; mammalian chromatin remodeling complex BRG1-associated factor 60C; mBAF60c; MGC111010; Rsc6p; Smarcd3; SWI/SNF complex 60 kDa subunit C; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3, isoform 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 3; Swp73-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6604, 66993 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6P9Z1, Q6STE5 | |
| SMARCD3 | |
| Recombinant protein encompassing a sequence within the center region of human SMARCD3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu