missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAFF/BLyS/TNFSF13B Antibody (belimumab), FITC, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals NBP3-27972F
355.30 EUR valable jusqu'au 2025-12-16
MEILLEUR PRIX promo! Utilisez le code promo "24090" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BAFF/BLyS/TNFSF13B Monoclonal antibody specifically detects BAFF/BLyS/TNFSF13B in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| BAFF/BLyS/TNFSF13B | |
| Monoclonal | |
| FITC | |
| ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4 | |
| TNFSF13B/ BAFF/ CD257 | |
| 0.1 mL | |
| Apoptosis, Cell Cycle and Replication, Cytokine Research, Immunology | |
| 10673 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| belimumab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu