Learn More
Band 3 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque:  Novus Biologicals NBP2-99575PE
Code nomenclature Nacres: NA.46
Code produit. 18651266
  • 407.00€ / 0.10mL
Description
Band 3 Polyclonal antibody specifically detects Band 3 in Rat samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Polyclonal
Immunohistochemistry-Paraffin
0.1 mL
Rat