Learn More
Invitrogen™ BCAS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA14398
Description
PA1-4398 detects NABC1 in human samples. PA1-4398 has been successfully used in Western blotting procedures. By Western blot, this antibody detects a ~60 kDa protein representing NABC1 in MCF-7 cell lysates. NABC1 can form dimers, which are readily detectable in MCF-7 cells. PA1-4398 immunizing peptide corresponds to amino acid residues 532-550 of human NABC1.
BCAS1 was identified through positional cloning and was found to be overexpressed in most but not all breast cancer cells lines. Little is known about this protein; BCAS1 is cytoplasmically localized and is thought to form homodimers through coiled-coil structures. High levels of BCAS1 mRNA are seen in brain and prostate tissues, with lower amounts observed in colon, intestine and testis. Cells engineered to overexpress BCAS1 did not lose anchorage-dependent growth or increase their rate of growth, suggesting that BCAS1 is not a prototypical oncogene. The BCAS1 gene was also found to be amplified in other carcinomas such as pancreatic carcinoma, suggesting that BCAS1 may play in important role in the control of cell proliferation.
Spécification
| BCAS1 | |
| Polyclonal | |
| Unconjugated | |
| BCAS1 | |
| 2210416M21Rik; 9030223A09Rik; AI841227; AIBC1; Amplified and overexpressed in breast cancer; amplified in breast cancer 1; band 83; Band83; BCAS1; breast carcinoma amplified sequence 1; breast carcinoma-amplified sequence 1; Breast carcinoma-amplified sequence 1 homolog; NABC1; Novel amplified in breast cancer 1; novel amplified in breast cancer 1 homolog; Pmes-2; Protein whose mRNA is enriched in synaptosomes 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8537 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O75363 | |
| BCAS1 | |
| Synthetic peptide: D(532) T N S L Q N G D K L Q K R P E K R Q(550). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.