missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCR Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22585-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene.Spécification
BCR | |
Polyclonal | |
Unconjugated | |
BCR | |
ALL, BCR, breakpoint cluster region, CML, D22S11, D22S662, PHL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
613 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11274 | |
BCR | |
BCR Fusion Protein Ag18281 | |
150 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |