missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDNF Mouse anti-Human, Rat, Clone: 1E5C5, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Spécification
| Antigène | BDNF |
|---|---|
| Clone | 1E5C5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
16838035
|
Proteintech
66292-1-IG-20UL |
20 μL |
145.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
16828035
|
Proteintech
66292-1-IG-150UL |
150 μL |
410.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer′s and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.Spécification
| BDNF | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Abrineurin, BDNF | |
| BDNF | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1E5C5 | |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Rat, Human | |
| P23363, P23560 | |
| 24225, 627 | |
| BDNF Fusion Protein Ag11329 | |
| Primary | |
| -20°C | |
| BDNF |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit