Learn More
Invitrogen™ BDNF Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA585730
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Cat (100%), Pig (100%), Chimpanzee (100%), Bovine (100%). Positive Control: mouse brain, rat brain, BDNF-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer's and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.
Spécification
| BDNF | |
| Polyclonal | |
| Unconjugated | |
| BDNF | |
| abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12064, 24225, 627 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.96 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P21237, P23363, P23560 | |
| BDNF | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human BDNF. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.