Learn More
beta Amyloid Antibody (bapineuzumab), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals NBP3-28816CL1
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| beta Amyloid | |
| Monoclonal | |
| CoraFluor 1 | |
| Abeta, Amyloid beta, beta-amyloid peptide | |
| Amyloid Beta | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction | |
| 351 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| bapineuzumab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |