missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Catenin Antibody (196624), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals FAB13292U
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Spécification
| beta-Catenin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 196624 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923 | |
| E. coli-derived recombinant human beta-Catenin, Ala2-Leu781, Accession # P35222 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 1499 | |
| Store at 4C in the dark. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu