Learn More
Beta Catenin Mouse, Clone: CTNNB1/1507, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 1499-MSM7-P1
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
| Beta Catenin | |
| Cell surface and cytoplasmic | |
| CTNNB1/1507 | |
| Unconjugated | |
| NP_001895 | |
| Mouse | |
| 92kDa | |
| Protein A/G Purified | |
| RUO | |
| 1499 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| CTNNB1 | |
| Recombinant human full-length beta-Catenin (p120) protein fragment | |
| HeLa or MCF-7 cells. Breast carcinoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |