missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-III Tubulin Antibody (TU-20), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB600-1018FR
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| beta-III Tubulin | |
| Monoclonal | |
| DyLight 680 | |
| Q13509 | |
| TUBB3 | |
| Peptide (C) 441-448 coupled to maleimide-activated keyhole limpet hemocyanin via cysteine added to the N-terminus of the neuron-specific peptide. | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers, Neuroscience, Phospho Specific | |
| 10381 | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| TU-20 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody TU-20 recognizes C-terminal peptide sequence ESESQGPK (aa 441-448) of neuron-specific human betaIII-tubulin. | |
| Store at 4C in the dark. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu