missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin Antibody (D66), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Marque: Novus Biologicals NB600-1514
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
beta Tubulin Monoclonal specifically detects beta Tubulin in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate, Rabbit samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Spécification
| beta Tubulin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 0.5-1 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
| TUBB | |
| Sea urchin (Lytechinus pictus) sperm axonemal proteins. | |
| 0.1 mL | |
| Breast Cancer | |
| 203068 | |
| Human, Mouse, Rat, Bovine, Canine, Chicken, Primate, Rabbit | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| D66 | |
| Unconjugated | |
| beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB600-1514 recognizes all mouse brain beta tubulin isoforms (beta and beta subpopulations), and sea urchin beta Tubulin. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu