Learn More
ACROBiosystems Biotinylated SARS-CoV-2 S protein RBD (N501Y), His, Avitag
Marque:  ACROBiosystems SPD-C82E6-25ug
Informations supplémentaires : Poids : 0.00010kg
Code produit. 17256540
  • 433.38€ / 25µg
Expédition estimée: 17-10-2022
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.2 kDa.
Spécification
Spécification
QHD43416.1 (N501Y)
28.2 kDa
Biotinylated SARS-CoV-2 S protein RBD (N501Y), His,Avitag (SPD-C82E6) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N501Y)).
-20°C
S Protein RBD
His Tag & Avi Tag
MABSol™ Biotin Labeled Proteins
Powder
25 μg
Arg 319 - Lys 537
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
Precision Avi
HEK293
SARS-CoV-2