missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLIMP1/PRDM1 Antibody (3H2-E8) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals (Bio-Techne) NB600-235JF635
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BLIMP1/PRDM1 Monoclonal antibody specifically detects BLIMP1/PRDM1 in Human,Mouse,Porcine samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),ChIP assay (ChIP),Immunocytochemistry/ Immunofluorescence
Spécification
| BLIMP1/PRDM1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PR-domain zinc finger protein 1 | |
| A fragment of mouse Blimp-1 corresponding to residues 199-409. [UniProt# Q60636] | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Cancer, Chromatin Research, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Zinc Finger | |
| 639 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ChIP Assay, Immunocytochemistry/Immunofluorescence | |
| 3H2-E8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu