missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bmf Monoclonal Antibody (OTI9B7)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA526878
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Bmf Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains a single BCL2 homology domain 3 (BH3), and has been shown to bind BCL2 proteins and function as an apoptotic activator. This protein is found to be sequestered to myosin V motors by its association with dynein light chain 2, which may be important for sensing intracellular damage and triggering apoptosis. Alternatively spliced transcript variants encoding different isoforms have been identified.
Spécification
| Bmf | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96LC9 | |
| BMF | |
| Full length human recombinant protein of BMF produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9B7 | |
| Unconjugated | |
| BMF | |
| AW260063; AW411872; Bcl2 modifying factor; Bcl-2 modifying factor; bcl-2-modifying factor; Bmf; FLJ00065 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 90427 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu