missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOK Recombinant Rabbit Monoclonal Antibody (6W4R6)
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA555399
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen Sequence: MEVLRRSSVF AAEIMDAFDR SPTDKELVAQ AKALGREYVH ARLLRAGLSW SAPERAAPVP GRLAEVCAVL LRLGDELEMI RPSVYRNVAR QLHISLQSEP.
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains all four BCL-2 like domains and is a pro-apoptotic BCL-2 protein identified in the ovary.
Spécification
| BOK | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O35425, Q792S6, Q9UMX3 | |
| Bok | |
| A synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6W4R6 | |
| Unconjugated | |
| Bok | |
| AI847676; apoptosis activator Mtd; AU021146; BCL2 family apoptosis regulator BOK; BCL2 related ovarian killer; BCL2L9; bcl2-L-9; Bcl-2-like protein 9; BCL2-related ovarian killer; bcl-2-related ovarian killer protein; BCL2-related ovarian killer protein; BOK; BOK, BCL2 family apoptosis regulator; Bok-BH3; BOKL; hBOK; matador; mtd; Protein matador | |
| Rabbit | |
| Protein A | |
| RUO | |
| 29884, 51800, 666 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction