Learn More
BPI Antibody (971526), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals FAB74681MFV610
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BPI Monoclonal antibody specifically detects BPI in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry
Spécification
| BPI | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry, Flow Cytometry | |
| 971526 | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry | |
| bactericidal permeability-increasing protein, bactericidal/permeability-increasing protein, CAP 57 | |
| Human embryonic kidney cell line HEK-293-derived transfected with human BPI, Val32-Lys487, Accession # P17213 | |
| RUO | |
| 671 | |
| Store at 4C in the dark. |