missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Mouse anti-Human, Clone: 9E10, TrueMAB™, Origene
Mouse Monoclonal Antibody
Marque: Origene Technologies TA150121S
This item is not returnable.
View return policy
Description
Synthetic peptide DYKDDDDK conjugated to KLH
c-Myc Monoclonal antibody specifically detects c-Myc in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
| c-Myc | |
| Monoclonal | |
| 0.64 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01106 | |
| Mouse | |
| Protein G | |
| RUO | |
| 4609 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 9E10 | |
| Unconjugated | |
| MYC | |
| AU016757; Avian myelocytomatosis viral (v-myc) oncogene homolog; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; mMyc; MRTL; Myc; Myc protein; Myc proto-oncogene protein; MYC proto-oncogene, bHLH transcription factor; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; myelocytomatosis viral oncogene homolog; Niard; Nird; Proto-oncogene c-Myc; RNCMYC; Transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene-like protein | |
| Synthetic peptide conjugated to KLH, corresponding to C terminal amino acids 408-432 of human c-Myc | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction