Learn More
C3a Receptor Monoclonal Antibody (74), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA517475
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
MA5-17475 targets C3a Receptor in FACS and IHC (F) applications and shows reactivity with Rat samples. The MA5-17475 immunogen is rBL-2H3 transfectants expressing rat C3aR.
C3AR1 is a receptor for the chemotactic and inflammatory peptide anaphylatoxin C3a. The receptor stimulates chemotaxis, granule enzyme release and superoxide anion production. (Uniprot)Spécification
C3a Receptor | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
O55197 | |
C3ar1 | |
RBL-2H3 transfectants expressing rat C3aR | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen) | |
74 | |
FITC | |
C3ar1 | |
anaphylatoxin C3a receptor; AZ3B; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; C3a-R; C3AR1; C3r1; complement component 3 receptor 1; complement component 3a receptor 1; HNFAG09 | |
Mouse | |
Protein G | |
RUO | |
84007 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |