Learn More
CAB39 Antibody (S03-4G7), Novus Biologicals™
Code produit 18618713 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μg
50 μg
Conditionnement:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18618713 100 μg 100µL
18697683 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18618713 Fournisseur Novus Biologicals Code fournisseur NBP319841100ul
Rabbit Monoclonal Antibody
CAB39 Monoclonal antibody specifically detects CAB39 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Antigène CAB39
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S03-4G7
Concentration 0.3 mg/mL
Conjugué Unconjugated
Dilution Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin
Formule 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Alias de gène calcium binding protein 39, calcium-binding protein 39, FLJ22682, MO25CGI-66, Protein Mo25
Espèces hôtes Rabbit
Immunogène A synthetic peptide of human CAB39 (Uniprot # Q9Y376)
Méthode de purification Affinity purified
Quantité 100 μg
État réglementaire RUO
Disciplines de recherche Autophagy, Cancer, Signal Transduction
Primaire ou secondaire Primary
Identification génétique (Entrez) 51719
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG