Learn More
Novus Biologicals™ CACNA1I Antibody, Novus Biologicals™
Code produit 30067205 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μg
25 μg
Conditionnement:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30067205 25 μg 25µg
30067159 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30067205 Fournisseur Novus Biologicals™ Code fournisseur NBP33082825ug
Rabbit Polyclonal Antibody
CACNA1I Polyclonal antibody specifically detects CACNA1I in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Tekniske data
Antigène CACNA1I
Applications Western Blot
Classification Polyclonal
Conjugué Unconjugated
Dilution Western Blot : 5-10 μg/mL
Formule Phosphate buffered saline
Alias de gène Ca(v)3.3, calcium channel, voltage-dependent, T type, alpha 1I subunit, voltage-dependent T-type calcium channel subunit alpha-1I, voltage-dependent, alpha 1I subunit, Voltage-gated calcium channel subunit alpha Cav3.3
Espèces hôtes Rabbit
Immunogène Rabbits were immunized with a synthetic peptide derived from the rat CACNA1I conjugated to KLH
Méthode de purification Ammonium sulfate precipitation
Quantité 25 μg
État réglementaire RUO
Disciplines de recherche Signal Transduction
Primaire ou secondaire Primary
Identification génétique (Entrez) 8911
Espèces cibles Human
Contenu et stockage Store at -20°C. Avoid freeze/thaw cycles.
Forme Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.