Learn More
Invitrogen™ CACNG2/4/8 Monoclonal Antibody (N245/36), APC

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA545584
Description
1 μg/mL of MA5-45584 was sufficient for detection of TARP Gamma2/4/8 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 35-55kDa. Does not cross-react with TARPGamma3. This antibody was formerly sold as clone S245-36.
Voltage-dependent calcium channel gamma-2 subunit, TARPGamma2 or Stargazin is encoded by the gene CACNG2 and is the transmembrane gamma subunit of the L-type calcium channel. Stargazin is in involved in regulation and trafficking of the ligand gated ion channel AMPA receptors. It is thought to play a role in learning and memory. Diseases associated with this gene include forms of deafness, Mental Retardation and childhood absnce epilepsy.
Spécification
| CACNG2/4/8 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4 | |
| O88602, Q71RJ2, Q8VHW2, Q8VHW5, Q8VHW9, Q8WXS5, Q9JJV4, Q9UBN1, Q9Y698 | |
| Mouse | |
| Protein G | |
| RUO | |
| 10369, 12300, 140725, 140729, 27092, 54377, 59283, 81905, 84347 | |
| 4°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| N245/36 | |
| APC | |
| Cacng8 | |
| Cacng2, Cacng4, Cacng8 | |
| Fusion protein amino acids 203-323 (Cytoplasmic C-terminus) of rat TARPGamma2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.