missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 1 Mouse anti-Human, Clone: CAPN1/1530, Novus Biologicals™
Mouse Monoclonal Antibody
249.00€
Spécification
| Antigène | Calpain 1 |
|---|---|
| Clone | CAPN1/1530 |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
Calpain 1 Monoclonal specifically detects Calpain 1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Spécification
| Calpain 1 | |
| 0.2 mg/ml | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| 10 mM PBS with 0.05% BSA | |
| 823 | |
| IgG1 κ | |
| Protein A or G purified |
| CAPN1/1530 | |
| ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Calcium-activated neutral proteinase 1, calpain 1, (mu/I) large subunit, Calpain mu-type, calpain, large polypeptide L1, calpain-1 catalytic subunit, Calpain-1 large subunit, CANP, CANP 1, CANPL1EC 3.4.22.52, Cell proliferation-inducing gene 30 protein, cell proliferation-inducing protein 30, EC 3.4.22, Micromolar-calpain, muCANPCANP1, muCL | |
| Recombinant full-length human Calpain 1 protein | |
| Primary | |
| Store at 4°C. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit