missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKI Polyclonal Antibody

Sheep Polyclonal Antibody
Marque: Invitrogen™ PA5149420
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
CAMK1 (CaMK1) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It is involved in cell differentiation, nervous system development, and signal transduction. CAMK1 is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.
Spécification
| CaMKI | |
| Polyclonal | |
| Unconjugated | |
| CAMK1 | |
| AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 8536 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q14012 | |
| CAMK1 | |
| E. coli-derived recombinant human CaM Kinase I alpha , Ser176-Arg254 (Ala224Ser). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu