missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Casein Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5119527
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Casein is the name for a family of related phosphoproteins. These proteins are commonly found in mammalian milk, making up 80% of the proteins in cow milk and between 20% and 45% of the proteins in human milk. Casein has a wide variety of uses, from being a major component of cheese, to use as a food additive, to a binder for safety matches. As a food source, casein supplies amino acids; carbohydrates; and two inorganic elements, calcium and phosphorus. Casein from bovine milk is a phosphoprotein. There are four main types of Casein which make up approximately 80% of the total protein in bovine milk.
Spécification
| Casein | |
| Polyclonal | |
| Unconjugated | |
| CSN1S1 | |
| AA959832; alpha S1-casein B; alpha-casein; Alpha-S1-casein; alpha-s1-casein precursor; Antioxidant peptide; AW212882; Bos d 8; CASA; casein alpha s1; casein alpha-S1; casein, alpha; Casoxin-D; Csn1; Csn1s1; Csna | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 282208 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 0.1% sodium azide | |
| P02662 | |
| CSN1S1 | |
| Bovine Casein. | |
| 200 μL | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu