missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Caveolin-1 Monoclonal antibody specifically detects Caveolin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
Specifications
| Antigène | Caveolin-1 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 2A7 |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA |
| Formule | PBS, pH 7.4, 150mM NaCl and 50% glycerol |
| Alias de gène | CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 |
| Espèces hôtes | Rabbit |
| Immunogène | A synthesized peptide derived from Human Caveolin-1 [UniProt Q03135] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?