Learn More
Caveolin-1 Antibody (2A7), Novus Biologicals™
Code produit 30043748 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μL
50 μL
Conditionnement:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30043748 50 μL 50µL
30043112 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30043748 Fournisseur Novus Biologicals Code fournisseur NBP32669550ul
Rabbit Monoclonal Antibody
Caveolin-1 Monoclonal antibody specifically detects Caveolin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Spécification
Antigène Caveolin-1
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 2A7
Conjugué Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA
Formule PBS, pH 7.4, 150mM NaCl and 50% glycerol
Alias de gène CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085
Espèces hôtes Rabbit
Immunogène A synthesized peptide derived from Human Caveolin-1 [UniProt Q03135]
Méthode de purification Affinity purified
Quantité 50 μL
État réglementaire RUO
Disciplines de recherche Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction
Primaire ou secondaire Primary
Identification génétique (Entrez) 857
Espèces cibles Human
Contenu et stockage Store at -20 to -70C. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG