missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBL, p Tyr700 Antibody (M161), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-23248
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CBL Monoclonal antibody specifically detects CBL in Human, Mouse, Rat samples. It is validated for Western Blot
Spécification
| CBL | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| M161 | |
| Western Blot 1:1000 | |
| Cas-Br-M (murine) ecotropic retroviral transforming sequence, Casitas B-lineage lymphoma proto-oncogene, CBL2E3 ubiquitin-protein ligase CBL, EC 6.3.2, EC 6.3.2.-, oncogene CBL2, Proto-oncogene c-Cbl, RING finger protein 55, RNF55C-CBL, Signal transduction protein CBL | |
| Clone (M161) was generated from a synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding tyrosine 700 in human c-Cbl. This sequence is highly conserved in rat and mouse c-Cbl. [UniProt# P22681] | |
| 0.1 mL | |
| Cancer, Phospho Specific, Signal Transduction, Zinc Finger | |
| 867 | |
| Store at -20°C. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu