missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL22/MDC Antibody (57203), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals FAB3361H
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CCL22/MDC Monoclonal antibody specifically detects CCL22/MDC in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Spécification
| CCL22/MDC | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 57203 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 | |
| E. coli-derived recombinant human CCL22/MDC, Gly25-Gln93, Accession # O00626.1 | |
| 0.5 mL | |
| Biologically Active Proteins | |
| 6367 | |
| Store at 4C in the dark. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu