Learn More
Invitrogen™ CCL5 (RANTES) Monoclonal Antibody (21418)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA523709
Description
When used in combination with the biotinylated anti-human CCL5/RANTES detection antibody in sandwich ELISAs, no significant cross-reactivity or interference is observed with recombinant human (rh) ANG, rhCNTF, rh beta -ECGF, rhEGF, mouse (m) EGF, rhEpo, bovine (b) FGF acidic, rhFGF acidic, bFGF basic, rhFGF basic, rhFGF-4, rhG-CSF, rhGM-CSF, recombinant mouse (rm) GM-CSF, rhGRO alpha, rhIFN-gamma, rhIGF-I, rhIGF-II, rhIL-1 alpha, rmIL-1 alpha, rhIL-1 beta, rmIL-1 beta, rhIL-1ra, recombinant rat (rr) IL-1ra, rhIL-2, rhIL-3, rmIL-3, rmIL-4, rhIL-5, rmIL-5, rhIL-6, rmIL-6, rhIL-7, rmIL-7, rhIL-8, rhIL-9, rmIL-9, rhIL-10, rhIL-11, rhLIF, rhMCP-1, rhM-CSF, rhMIP-1 alpha, rmMIP-1 alpha, rhMIP-1 beta, rmMIP-1 beta, rhOSM, human (h) PDGF, pPDGF, rhPDGF-AA, rhPDGF-AB, rhPDGF-BB, rhSCF, rmSCF, rhSLPI, rhTGF-alpha, pTGF-beta 1, hTGF-beta 1, rhTGF-beta 1, pTGF-beta 2, rhTGF-beta 2, recombinant canine TGF-beta 3, rhTGF-beta 3, recombinant amphibian TGF-beta 5, rhTNF-alpha, rmTNF-alpha, or rmTNF-beta. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CCL5 (RANTES) is a chemokine gene located on the q-arm of chromosome 17. Chemokines are a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. CCL5, a member of the CC subfamily, functions as a chemoattractant for blood monocytes, memory T helper cells, and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor chemokine (C-C motif) receptor 5 (CCR5), and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. Alternative splicing results in multiple transcript variants that encode different isoforms. Diseases associated with CCL5 include Human Immunodeficiency Virus Type 1 and Periapical Granuloma.
Spécification
| CCL5 (RANTES) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P13501 | |
| CCL5 | |
| E. coli-derived recombinant human CCL5/RANTES. | |
| 500 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| ELISA, Neutralization, Western Blot | |
| 21418 | |
| Unconjugated | |
| CCL5 | |
| beta-chemokine RANTES; CC chemokine; C-C motif chemokine 5; C-C motif chemokine ligand 5; CCL5; chemokine (C-C motif) ligand 5; D17S136E; EoCP; eosinophil chemotactic cytokine; MGC17164; MuRantes; OTTHUMP00000197106; RANTES; RANTES protein; RANTES(3-68); RANTES(4-68); regulated upon activation normal T-cell expressed and secreted; regulated upon activation, normally T-expressed, and presumably secreted; Scya5; SIS δ; SISd; SIS-delta; SISδ; small inducible cytokine A5; small inducible cytokine subfamily A (Cys-Cys), member 5; small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; Unknown (protein for MGC:127014) | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6352 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.