Learn More
CCR5 Antibody (CTC5R), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals FAB1802RA
Code nomenclature Nacres: NA.46
488.75 EUR valable jusqu'au 2025-12-16
MEILLEUR PRIX promo! Utilisez le code promo "24090" pour bénéficier de cette offre.
Code produit. 18629493
  • 569.00€ / 0.10mL
Définissez la recherche en immunologie avec vos termes
Utilisez le code promo 24090
Afficher toutes les promotions
Description
CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Spécification
Spécification
CCR5
Monoclonal
APC
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
CTC5R
Flow Cytometry, CyTOF-ready
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
CHO Chinese hamster ovary cell line transfected with human CCR5
0.1 mL
Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Signal Transduction
1234
Store at 4°C in the dark.
IgG1