missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCRL2 Mouse anti-Human, Clone: 2C1E11, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66611-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CCRL2 (C-C chemokine receptor-like 2), also known as chemokine receptor X, human chemokine receptor (HCR), CRAM-A or CRAM-B, is a seven pass transmembrane protein expressed by monocytes, neutrophils, and dendritic cells of immunal tissues (predomily spleen, fetal liver, bone marrow and lymph node). It shares over 40% homology with other C-C chemokine receptors but does not share the conserved DRY motif which is among the important motifs necessary for signalling and ligand-binding. C-C chemokine receptors are G-protein coupled, seven pass transmembrane domain proteins whose major physiological role is to function in the chemotaxis of T cells and phagocytic cells to areas of inflammation.Spécification
| CCRL2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00421 | |
| CCRL2 | |
| CCRL2 Fusion Protein Ag4045 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2C1E11 | |
| Unconjugated | |
| CCRL2 | |
| C C chemokine receptor like 2, CCR11, CCR6, CCRL2, Chemokine receptor CCR11, Chemokine receptor X, CKRX, CRAM, CRAM A, CRAM B, HCR | |
| Mouse | |
| Protein G | |
| RUO | |
| 9034 | |
| -20°C | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu