Learn More
CCSAP Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28644-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CCSAP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CCSAP. This antibody recognizes mouse, rat antigen. The CCSAP antibody has been validated for the following applications: WB, ELISA analysis.Spécification
| CCSAP | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 126731 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 220 μg/mL | |
| FLJ37296; FLJ41471 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Mouse, Rat | |
| 30-36 kDa | |
| Liquid |