Learn More
Abnova™ CCT8 Recombinant Protein
Human CCT8 full-length ORF ( AAH05220, 1 a.a. - 27 a.a.) recombinant protein with GST-tag at N-terminal
Marque: Abnova™ H00010694-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Molecular weight: 28.71kDa
- Preparation method: in vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8 in the elution buffer
- Quality Control Testing: 12.5% SDS-PAGE stained with Coomassie Blue
Sequence:
MALHVPKAPGFAQMLKEGAKVRAEGKE
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spécification
AAH05220 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
28.71 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MALHVPKAPGFAQMLKEGAKVRAEGKE | |
C21orf112/Cctq/D21S246/KIAA0002/PRED71 | |
CCT8 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
10694 | |
CCT8 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCT8 | |
Human | |
Recombinant | |
Solution |