Learn More
CD103 Mouse anti-Human, Brilliant Violet 786, Clone: Ber-ACT8, BD Optibuild™
Mouse Monoclonal Antibody
Marque: BD Biosciences 743654
Description
The Ber-ACT8 monoclonal antibody specifically binds to the human mucosal lymphocyte antigen 1 (HML-1). This is a 175 kDa type I transmembrane glycoprotein, also known as integrinαE (ITGAE, Integrin alpha-E) and CD103. It is found on >90% of intestinal intraepithelial lymphocytes (iIEL) associated with integrinβ7. CD103 is also expressed on lamina propria T lymphocytes in the intestine and on phytohemagglutinin-stimulated peripheral blood lymphocytes. It is rarely expressed on resting peripheral blood lymphocytes. It has been suggested that CD103 may have an accessory function for activation of iIEL. CD103 has been reported as a useful tool in hairy cell leukemia research.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
Spécification
| CD103 | |
| Ber-ACT8 | |
| Brilliant Violet 786 | |
| ITGAE, Integrin alpha-E, ITAE, HML-1 antigen, Mucosal lymphocyte 1, HUMINAE | |
| Human MAPS16 T Cell Line | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.