Learn More
CD105 (Endoglin) Mouse, anti-Human, APC, Clone: 43A3, STEMCELL Technologies
Mouse Monoclonal Antibody
Marque:  Stemcell Technologies 60039AZ.1
Encompass_Preferred
Code nomenclature Nacres: NA.46
Code produit. 17148711
  • 276.75€ / 25 tests
Description
  • The 43A3 antibody reacts with CD105 (endoglin), an ∼180 kDa cell surface glycoprotein which is a disulfide-bonded homodimer of ∼90 kDa type I transmembrane subunits
  • CD105 is a component of the TGF-β receptor complex and is expressed by vascular endothelial smooth muscle cells, syncytiotrophoblasts of placenta and activated macrophages, and at relatively low levels by stromal fibroblasts
  • Its expression is also observed in some types of tumors, and levels are up-regulated on the endothelium during angiogenesis
  • CD105 has important roles in angiogenesis, cardiovascular development, and vascular remodeling, and the protein serves a regulatory role in cytoskeletal reorganization by modulating the sites of focal adhesion and cellular migration
  • This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Spécification
Spécification
CD105 (Endoglin)
Monoclonal
APC
25 Tests
Human, Mouse
Hematopoietic Stem and Progenitor Cells, Mesenchymal Stem and Progenitor Cells
Immunoprecipitation, Western Blot, Flow Cytometry
43A3
Mouse
Primary
IgG1 κ
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.