missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD115 (CSF-1R) Rat anti-Mouse , APC, Clone: T38-320, BD Biosciences™
Rat Monoclonal Antibody
Marque: BD Biosciences 567027
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD115 (CSF-1R) Monoclonal antibody specifically detects CD115 (CSF-1R) in Mouse samples. It is validated for Flow Cytometry
Spécification
| CD115 (CSF-1R) | |
| Monoclonal | |
| APC | |
| M-CSFR; M-CSF-R; CSF-1 Receptor; CSF-1R; Csf1r; Csfmr; Fms; c-Fms; Fim-2 | |
| Mouse CD115 Recombinant Protein | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| T38-320 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu