missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11a/Integrin alpha-L Mouse anti-Human, Clone: 1E3H12, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66256-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD11a (LFA-1 alpha, ITGAL) together with CD18 constitute leukocyte function-associated antigen 1 (LFA-1), the alphaLbeta2 integrin. CD11a is implicated in activation of LFA-1 complex. CD11a plays a central role in leukocyte intercellular adhesion through interactions with its ligands, ICAMs 1-3 (intercellular adhesion molecules 1 through 3), and also functions in lymphocyte costimulatory signaling. CD11a is expressed on the plasma membrane of leukocytes in a low-affinity conformation. Cell stimulation by chemokines or other signals leads to induction the high-affinity conformation, which supports tight binding of CD11a to its ligands, the intercellular adhesion molecules ICAM-1, -2, -3. CD11a is thus involved in interaction of various immune cells and in their tissue-specific settlement, but participates also in control of cell differentiation and proliferation and of T-cell effector functions. Blocking of CD11a function by specific antibodies or small molecules has become an important therapeutic approach in treatment of multiple inflammatory diseases. For example, humanized anti- CD11a antibody Efalizumab (Raptiva) is being used to interfere with T cell migration to sites of inflammation, binding of cholesterol-lowering drug simvastatin to CD11a allosteric site leads to immunomodulation and increase in lymphocytic cholinergic activity. Two transcript variants encoding different isoforms of CD11a have been found. Diseases associated with CD11a dysfunction include benign cephalic histiocytosis and leukocyte adhesion deficiency.Spécification
| CD11a/Integrin alpha-L | |
| Monoclonal | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20701 | |
| ITGAL | |
| CD11a/Integrin alpha-L Fusion Protein Ag7813 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 1E3H12 | |
| Unconjugated | |
| ITGAL | |
| CD11A, CD11a/Integrin alpha-L, Integrin alpha L, ITGAL, LFA 1, LFA 1A, LFA1A | |
| Mouse | |
| Protein G | |
| RUO | |
| 3683 | |
| -20°C | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu