missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11b Antibody (M1/70.15), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Marque: Novus Biologicals NB600-1327JF646
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD11b Monoclonal antibody specifically detects CD11b in Human, Mouse, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Spécification
| CD11b | |
| Monoclonal | |
| Janelia Fluor 646 | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
| Rat Monoclonal CD11b Antibody (M1/70.15) was made to T cell enriched splenocytes from B10 mice. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Biology, Cellular Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 3684 | |
| Store at 4°C in the dark. | |
| IgG2b λ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| M1/70.15 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rabbit | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu