missing translation for 'onlineSavingsMsg'
Learn More

CD137 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody

Marque:  Invitrogen 500-P167GBT-50UG

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17893903

  • 239.00€ / 50µg
Expédition estimée: 12-10-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

AA Sequence of recombinant protein: MERTRSLQDP CSNCPAGTFC DNNRNQICSP CPPNSFSSAG GQRTCDICRQ CKGVFRTRKE CSSTSNAECD CTPGFHCLGA GCSMCEQDCK QGQELTKKGC KNCCFGTFND QKRGICRPWT NCSLDGKSVL VNGTKERDVV CGPSPADLSP GASSVTPPAP AREPGHS. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human 4-1BB Receptor. Anti-Human 4-1BB Receptor-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect h4-1BB Receptor by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human 4-1BB Receptor (500-P167G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant h4-1BB Receptor. Western Blot: To detect h4-1BB Receptor by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant h4-1BB Receptor is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

CD137 Polyclonal antibody specifically detects CD137 in Human samples. It is validated for ELISA, Western Blot
Spécification

Spécification

CD137
Polyclonal
PBS with no preservative
Q07011
Goat
Antigen affinity chromatography
RUO
3604
-20°C
ELISA, Western Blot
Biotin
TNFRSF9
4 1 BB; 41BB; 4-1BB; 4-1BB ligand receptor; A930040I11Rik; AA408498; AI325004; Cd137; CD137 antigen; CDw137; EGK_00183; homolog of mouse 4-1BB; ILA; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; Ly63; receptor protein 4-1BB; s4 1 BB; s4 1BB; s41BB; sCD137; secreted CD137 antigen; soluble 4 1 BB; soluble 4 1BB; soluble 41BB; soluble CD137; T cell antigen ILA; T-cell antigen 4-1BB; T-cell antigen 4-1BB homolog; T-cell antigen ILA; TNF receptor superfamily member 9; TNFRSF9; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9
E.coli-derived Recombinant Human 4-1BB Receptor
50 μg
Primary
Human
Lyophilized
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions