Learn More
Invitrogen™ CD142 Monoclonal Antibody (HTF-1), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
3305.00€ - 14910.00€
Spécification
| Antigène | CD142 |
|---|---|
| Clone | HTF-1 |
| Concentration | 1 mg/mL |
| Classification | Monoclonal |
| Conjugué | Functional Grade |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
17121152
|
Invitrogen™
16-1429-38 |
5 mg |
3305.00€
5mg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
17101152
|
Affymetrix eBioscience
16-1429-025 |
25 mg | |||||||
|
17111152
|
Affymetrix eBioscience
16-1429-050 |
50 mg | |||||||
Description
This HTF-1 MAb reacts with human CD142, which is also known as Tissue Factor. Expression of this type I transmembrane glycoprotein on endothelial cells, monocytes, macrophages, and platelets can be induced by inflammatory mediators (e.g., LPS, IL-1b, TNFa, PMA, or endotoxin). On the other hand, CD142 is expressed constitutively by some tumor cells (e.g., lung, pancreatic, breast, and colon) and non-immune tissues such as the vasculature, central nervous system, kidney, epithelia, and placenta. Studies have also suggested that CD142 exists as a soluble form that circulates in blood. CD142 initiates blood coagulation by associating with and activating the circulating factors VII and VIIa. The HTF-1 antibody has been reported to exhibit blocking activity.
This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII. The resulting complex provides a catalytic event that is responsible for initiation of the coagulation protease cascades by specific limited proteolysis. Unlike the other cofactors of these protease cascades, which circulate as nonfunctional precursors, this factor is a potent initiator that is fully functional when expressed on cell surfaces. There are 3 distinct domains of this factor: extracellular, transmembrane, and cytoplasmic. This protein is the only one in the coagulation pathway for which a congenital deficiency has not been described.Spécification
| CD142 | |
| 1 mg/mL | |
| Functional Grade | |
| Mouse | |
| Human | |
| P13726 | |
| 2152 | |
| Primary | |
| 4°C | |
| F3 |
| HTF-1 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| AA409063; CD142; Cf3; Cf-3; Coagulation factor III; coagulation factor III (thromboplastin, tissue factor); coagulation factor III, tissue factor; F3; TF; TFA; Thromboplastin; Tissue factor | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.