Learn More
CD154 Armenian Hamster, PE, Clone: MR1, enQuireBio™
Armenian Hamster Monoclonal Antibody
Marque:  EnquireBio QAB88-PE-25ug
Code nomenclature Nacres: NA.46
Code produit. 15982598
  • 131.25€ / 25µg
Spécification
CD154
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Cd40lg
Protein A/G Purified
RUO
21947
IgG
Flow Cytometry
MR1
PE
NP_035746
Armenian Hamster
25 μg
Primary
Store at 2-4°C.