missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD162 Monoclonal Antibody (TC2), APC, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA110115
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.Spécification
CD162 | |
Monoclonal | |
APC | |
SELPLG | |
CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
6404 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
TC2 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q14242 | |
SELPLG | |
Human thymocytes | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |