Learn More
Invitrogen™ CD162 (PSGL-1) Monoclonal Antibody (4RA10), Alexa Fluor™ 647, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
204.00€ - 465.00€
Spécification
| Antigène | CD162 (PSGL-1) |
|---|---|
| Clone | 4RA10 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
16676384
|
Invitrogen™
51-1621-80 |
25 μg |
204.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16686384
![]() |
Invitrogen™
51-1621-82 |
100 μg |
465.00€
1 pièce |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Description: This 4RA10 monoclonal antibody reacts with mouse CD162, which is also known as P-selectin glycoprotein ligand-1 (PSGL-1). This clone has been successfully used in flow cytometry and has been reported to block binding of CD162 to CD62P and CD62L. Applications Reported: This 4RA10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4RA10 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser
CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.Spécification
| CD162 (PSGL-1) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG | |
| SELPLG | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| 4RA10 | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Rat | |
| Mouse | |
| Q62170 | |
| 20345 | |
| Mouse PSGL-1 human IgG1 fusion protein. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| SELPLG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.