missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD18 Mouse anti-Human, BUV661, Clone: L130, BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 750750
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD18 Monoclonal antibody specifically detects CD18 in Human samples. It is validated for Flow Cytometry
Spécification
| CD18 | |
| Monoclonal | |
| Brilliant Ultraviolet 661 | |
| ITGB2; Integrin beta-2; Beta subunit for LFA-1; MAC-1; p150 95; or aDb2 | |
| Thymic TCR γδ+ T lymphocytes | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| L130 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu