Learn More
CD184 (CXCR4) Monoclonal Antibody (12G5), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marque: Affymetrix eBioscience 16-9999-025
Description
The 12G5 MAb reacts with human CD184 (CXCR4), also termed Fusin, LESTR, or HUMSTR. CD184 is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. CD184 is expressed predominantly on naive T cell subsets of peripheral blood and is rapidly upregulated by PHA and IL-2 stimulation. The 12G5 antibody shows partial inhibition of chemotaxis and calcium influx induced by SDF-1 (the natural ligand of CD184), blocks CD4-independent HIV-2 infection, and blocks CD4-dependent infection by some T-tropic HIV-1 isolates.
CD184 (CXCR4) Monoclonal antibody specifically detects CD184 (CXCR4) in Human samples. It is validated for Flow Cytometry, Functional Assay, Neutralization
Spécification
| CD184 (CXCR4) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P61073 | |
| Mouse | |
| 25 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry, Functional Assay, Neutralization | |
| 12G5 | |
| Functional Grade | |
| Cxcr4 | |
| b2b220Clo; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD antigen CD184; CD184; CD184 antigen; chemokine (C-X-C motif) receptor 4; chemokine (C-X-C) receptor 4; chemokine receptor; chemokine receptor (LCR1); chemokine receptor 4; chemokine receptor CXCR4; Cmkar4; CXC; CXC chemokine receptor; C-X-C chemokine receptor type 4; CXC motif chemokine; C-X-C motif chemokine receptor 4; Cxcr4; CXC-R4; CXCR-4; D2S201E; FB22; Fusin; HM89; HSY3RR; HUMSTR; LAP3; LAP-3; LCR1; LESTR; leukocyte-derived seven transmembrane domain receptor; leukocyte-expressed seven-transmembrane-domain; lipopolysaccharide-associated protein 3; LPS-associated protein 3; neuropeptide Y receptor Y3; neuropeptide Y3 receptor; NPY3R; NPYR; NPYRL; NPYY3R; PB-CKR; PBSF/SDF-1; Pre-B-cell-derived chemokine receptor; SDF-1 receptor; Sdf1r; seven transmembrane helix receptor; seven-transmembrane-segment receptor, spleen; stromal cell-derived factor 1 receptor; WHIM; WHIMS | |
| Affinity chromatography | |
| RUO | |
| 7852 | |
| 4° C | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.