missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1c Monoclonal Antibody (L161), NovaFluor™ Red 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
191.00€ - 406.00€
Spécification
| Antigène | CD1c |
|---|---|
| Clone | L161 |
| Concentration | 0.1 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
This gene encodes a member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail, and requires vesicular acidification to bind lipid antigens.Spécification
| CD1c | |
| 0.1 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| BDCA1; canCD1c; CD1; CD1A; CD1C; CD1C antigen, c polypeptide; CD1c molecule; cortical thymocyte antigen CD1C; differentiation antigen CD1-alpha-3; R7; RP11-101J8.3; T-cell surface glycoprotein CD1c | |
| CD1C | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
| L161 | |
| Flow Cytometry | |
| NovaFluor Red 755 | |
| Mouse | |
| Human | |
| P29017 | |
| 911 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD1C |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit