Learn More
CD1d Rat anti-Mouse, FITC, Clone: 1B1, BD
Rat Monoclonal Antibody
153.01€ - 296.39€
Spécification
| Antigène | CD1d |
|---|---|
| Clone | 1B1 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
15811309
|
BD Biosciences
561756 |
50 μg |
154.00€ 153.01€ / 1 pièce Économisez 0.99€ 0.64% de réduction |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
15877418
|
BD Biosciences
553845 |
0.5 mg |
297.00€ 296.39€ / 1 pièce Économisez 0.61€ 0.21% de réduction |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
The 1B1 monoclonal antibody specifically binds to mouse CD1d, a 48-kDa glycoprotein with structural homology to the major histocompatibility complex (MHC) class I molecules. The structure, expression, and functions of CD1 antigens are complex and have been reviewed. mAb 1B1 detects CD1d at varying levels on most types of bone marrow and peripheral leukocytes and on epithelial, dendritic, and lymphoid cells in the thymus. It appears to recognize CD1d only in association with β2m. CD1d has been reported to be expressed by gastrointestinal tract epithelium and in the cytoplasm of hepatocytes as detected via immunohistochemical staining of frozen sections with mAb 3C11 (Cat. No. 559871, for the purified antibody), suggesting a possible role for CD1d in mucosal immunity. However, CD1d expression was not detectable, via flow cytometry, on intestinal epithelial cells in studies using the anti-CD1d mAbs 3C11, 1B1, and 9C7. The 1B1 antibody competes with mAb 3C11 in binding to mouse splenocytes.
Spécification
| CD1d | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| IgG2b κ | |
| Affinity Purified | |
| Cd1d1; Cd1.1; Cd1a; Cd1d; Ly-38 |
| 1B1 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.