Learn More
CD201 (EPCR) Monoclonal Antibody (eBio1560 (1560)), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
8960.00€ - 14910.00€
Spécification
| Antigène | CD201 (EPCR) |
|---|---|
| Clone | eBio1560 (1560) |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry, Neutralization |
| Classification | Monoclonal |
Description
The eBio1560 MAb reacts with mouse Endothelial Protein C Receptor (EPCR, CD201), a 25 kDa Type 1 transmembrane protein expressed on endothelial cells. EPCR exhibits sequence and structural homology with the MHC class I/CD1 family of proteins. EPCR is a ligand for Protein C and plays an important role in augmenting Protein C activation by the thrombin-thrombomodulin complex and in regulating blood coagulation and inflammation. Deletion of EPCR results in embryonic lethality, at least partically due to placental thrombosis. Recently, it was demonstrated that EPCR expression identified cells in the bone marrow that are capable of hematopoietic reconstitution activity comparable to hematopoietic stem cells isolated with conventional methods. The eBio1560 MAb can be used for the detection of these hematopoietic stem cells, however the eBiomRCR-16 MAb should only be used for the detection of CD201 on endothelial cells.
CD201 (EPCR) Monoclonal antibody specifically detects CD201 (EPCR) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry, NeutralizationSpécification
| CD201 (EPCR) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| activated protein C receptor; AI325044; APC receptor; bA42O4.2; CCCA; CCD41; CD antigen CD201; CD201; CD201 antigen; cell cycle, centrosome-associated protein; Centrocyclin; Centrosomal protein CCD41; endothelial cell protein C receptor; Endothelial protein C receptor; Epcr; PROCR; protein C receptor; protein C receptor, endothelial | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
| eBio1560 (1560) | |
| Flow Cytometry, Functional Assay, Immunohistochemistry, Neutralization | |
| Functional Grade | |
| Rat | |
| Mouse | |
| Q64695 | |
| 19124 | |
| Primary | |
| 4° C | |
| Procr |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.