Learn More
CD207 (Langerin) Monoclonal Antibody (eBioRMUL.2), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marque: Affymetrix eBioscience 14-2073-95
Description
RMUL.2 is a MAb that reacts with the predicted extracellular domain of mouse CD207, also known as Langerin. Mouse CD207 is expressed in a distinct subset of dendritic cells called Langerhans cells (LC). Mouse CD207 is a 48 kDa, C-type lectin and putative transmembrane protein that likely plays a role in antigen recognition and uptake. LC are located in the epidermis and upon activation reduce expression of CD207 and begin migrating through the dermis towards lymphatic vessels. Expression of mouse CD207 in LC has been correlated to the presence of Birbeck Granules. Furthermore, the formation of Birbeck Granules has been observed upon transfection of CD207 cDNA into fibroblast cell lines. Western Blotting with RMUL.2 reveals a 48 kDa band in lysate from mouse ear epidermis as well as in lysates from cells transfected with mouse CD207 cDNA. Cross reactivity of this antibody with other proteins has not been determined. Preliminary data from mouse CD207 cDNA transfected cells suggests that RMUL.2 can be used for intracellular staining followed by flow cytometric analysis. We have observed in our transfectants the majority of protein is expressed intracellularly with a small percentage on the cell surface. In addition, RMUL.2 has also been tested by immunohistology on mouse ear epidermal sheets and shows specific staining.
CD207 (Langerin) Monoclonal antibody specifically detects CD207 (Langerin) in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Spécification
| CD207 (Langerin) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q8VBX4 | |
| Rat | |
| 10 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| eBioRMUL.2 | |
| Unconjugated | |
| CD207 | |
| CD 207 antigen; Cd207; CD207 antigen; CD207 antigen, langerin; CD207 molecule; CD207 molecule, langerin; Clec4k; C-type lectin domain family 4 member K; C-type lectin domain family 4, member K; Langerhans cell specific c-type lectin; langerin; RGD1565913 | |
| Affinity chromatography | |
| RUO | |
| 246278 | |
| 4° C | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.